Learn More
Biomatik Corporation KYNU anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier: Biomatik Corporation CAC07646
Kynu Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KYNU. The Kynu Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Kynureninase Protein (99-214Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: KYNU. Accession Number: Q16719. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.