Learn More
Biomatik Corporation C6orf10 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07648
Encompass_Preferred
The C6orf10 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Uncharacterized protein C6orf10 protein (239-476AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:2000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Uncharacterized protein C6orf10, C6orf10. UniProt: Q5SRN2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7233