Learn More
Biomatik Corporation RABGAP1L anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507238
Encompass_Preferred
Rabgap1L Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RABGAP1L. The Rabgap1L Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Rab Gtpase-Activating Protein 1-Like, Isoform 10 Protein (11-262Aa). Applications: ELISA, WB, IHC. Target Synonyms: RABGAP1L. Accession Number: B7ZAP0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7238 Supplier Biomatik Corporation Supplier No. CAC07653
Rabgap1L Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RABGAP1L. The Rabgap1L Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Rab Gtpase-Activating Protein 1-Like, Isoform 10 Protein (11-262Aa). Applications: ELISA, WB, IHC. Target Synonyms: RABGAP1L. Accession Number: B7ZAP0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.