Learn More
Biomatik Corporation CARM1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07657
Encompass_Preferred
The CARM1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Histone-arginine methyltransferase CARM1 protein (209-379AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:2000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Histone-arginine methyltransferase CARM1, Coactivator-associated arginine methyltransferase 1, Protein arginine N-methyltransferase 4, CARM1, PRMT4. UniProt: Q86X55. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7242