Learn More
Biomatik Corporation ZBTB46 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07661
Encompass_Preferred
Zbtb46 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ZBTB46. The Zbtb46 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Zinc Finger And Btb Domain-Containing Protein 46 Protein (174-362Aa). Applications: ELISA, WB, IHC. Target Synonyms: ZBTB46. Accession Number: Q86UZ6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7246