Learn More
Biomatik Corporation P3H2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507248
Encompass_Preferred
P3H2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to P3H2. The P3H2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Prolyl 3-Hydroxylase 2 Protein (301-528Aa). Applications: ELISA, WB, IHC. Target Synonyms: P3H2. Accession Number: Q8IVL5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7248 Supplier Biomatik Corporation Supplier No. CAC07663
P3H2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to P3H2. The P3H2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Prolyl 3-Hydroxylase 2 Protein (301-528Aa). Applications: ELISA, WB, IHC. Target Synonyms: P3H2. Accession Number: Q8IVL5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.