Learn More
Biomatik Corporation SLC38A9 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier: Biomatik Corporation CAC07667
Slc38A9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC38A9. The Slc38A9 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sodium-Coupled Neutral Amino Acid Transporter 9 Protein (1-119Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC38A9. Accession Number: Q8NBW4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.