Learn More
Biomatik Corporation FBXW5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07669
Encompass_Preferred
The FBXW5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human F-box/WD repeat-containing protein 5 protein (265-494AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: F-box/WD repeat-containing protein 5, F-box and WD-40 domain-containing protein 5, FBXW5, FBW5, PP3971. UniProt: Q969U6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7254