Learn More
Biomatik Corporation ARHGDIG anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507258
Encompass_Preferred
The ARHGDIG Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Rho GDP-dissociation inhibitor 3 protein (1-225AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Rho GDP-dissociation inhibitor 3Rho GDI 3, Rho-GDI gamma, ARHGDIG. UniProt: Q99819. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7258 Supplier Biomatik Corporation Supplier No. CAC07673
The ARHGDIG Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Rho GDP-dissociation inhibitor 3 protein (1-225AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Rho GDP-dissociation inhibitor 3Rho GDI 3, Rho-GDI gamma, ARHGDIG. UniProt: Q99819. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.