Learn More
Biomatik Corporation SCPEP1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07677
Encompass_Preferred
Scpep1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SCPEP1. The Scpep1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Retinoid-Inducible Serine Carboxypeptidase Protein (201-452Aa). Applications: ELISA, WB, IHC. Target Synonyms: SCPEP1. Accession Number: Q9HB40. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7262