Learn More
Biomatik Corporation ABCG8 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507265
Encompass_Preferred
The ABCG8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human ATP-binding cassette sub-family G member 8 protein (176-413AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:2000-1:10000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: ATP-binding cassette sub-family G member 8, Sterolin-2, ABCG8. UniProt: Q9H221. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7265 Supplier Biomatik Corporation Supplier No. CAC07680
The ABCG8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human ATP-binding cassette sub-family G member 8 protein (176-413AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:2000-1:10000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: ATP-binding cassette sub-family G member 8, Sterolin-2, ABCG8. UniProt: Q9H221. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.