Learn More
Biomatik Corporation BTNL2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507268
Encompass_Preferred
The BTNL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Butyrophilin-like protein 2 protein (26-202AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:2000-1:10000, IHC:1:20-1:200. Alternative Names: Butyrophilin-like protein 2BTL-II, BTNL2. UniProt: Q9UIR0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7268 Supplier Biomatik Corporation Supplier No. CAC07683
The BTNL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Butyrophilin-like protein 2 protein (26-202AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:2000-1:10000, IHC:1:20-1:200. Alternative Names: Butyrophilin-like protein 2BTL-II, BTNL2. UniProt: Q9UIR0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.