Learn More
Biomatik Corporation DHX33 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507270
Encompass_Preferred
The DHX33 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Putative ATP-dependent RNA helicase DHX33 protein (116-357AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Putative ATP-dependent RNA helicase DHX33, DEAH box protein 33, DHX33, DDX33. UniProt: Q9H6R0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7270 Supplier Biomatik Corporation Supplier No. CAC07685
The DHX33 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Putative ATP-dependent RNA helicase DHX33 protein (116-357AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Putative ATP-dependent RNA helicase DHX33, DEAH box protein 33, DHX33, DDX33. UniProt: Q9H6R0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.