Learn More
Biomatik Corporation PPM1H anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507273
Encompass_Preferred
Ppm1H Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PPM1H. The Ppm1H Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Phosphatase 1H Protein (73-289Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PPM1H. Accession Number: Q9ULR3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7273 Supplier Biomatik Corporation Supplier No. CAC07688
Ppm1H Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PPM1H. The Ppm1H Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Phosphatase 1H Protein (73-289Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PPM1H. Accession Number: Q9ULR3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.