Learn More
Biomatik Corporation ICE1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07690
Encompass_Preferred
Ice1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ICE1. The Ice1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Little Elongation Complex Subunit 1 Protein (1213-1505Aa). Applications: ELISA, WB, IHC. Target Synonyms: ICE1. Accession Number: Q9Y2F5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7275