Learn More
Biomatik Corporation ASS1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07696
Encompass_Preferred
The ASS1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Argininosuccinate synthase protein (168-222AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Argininosuccinate synthase, Citrulline--aspartate ligase, ASS1, ASS. UniProt: P00966. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7281