Learn More
Biomatik Corporation CHAT anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507286
Encompass_Preferred
The CHAT Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Choline O-acetyltransferase protein (354-592AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Choline O-acetyltransferase, CHAT. UniProt: P28329. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7286 Supplier Biomatik Corporation Supplier No. CAC07701
The CHAT Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Choline O-acetyltransferase protein (354-592AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Choline O-acetyltransferase, CHAT. UniProt: P28329. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.