Learn More
Biomatik Corporation DDX6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07702
Encompass_Preferred
The DDX6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Probable ATP-dependent RNA helicase DDX6 protein (1-149AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Probable ATP-dependent RNA helicase DDX6Curated, ATP-dependent RNA helicase p54, DEAD box protein 6, Oncogene RCK, DDX6, HLR2, RCK. UniProt: P26196. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7287