Learn More
Biomatik Corporation EIF4A3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507289
Encompass_Preferred
The EIF4A3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Eukaryotic initiation factor 4A-III protein (6-166AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Eukaryotic initiation factor 4A-III, ATP-dependent RNA helicase DDX48, ATP-dependent RNA helicase eIF4A-3, DEAD box protein 48, Eukaryotic initiation facto, EIF4A3, DDX48, KIAA0111. UniProt: P38919. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7289 Supplier Biomatik Corporation Supplier No. CAC07704
The EIF4A3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Eukaryotic initiation factor 4A-III protein (6-166AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Eukaryotic initiation factor 4A-III, ATP-dependent RNA helicase DDX48, ATP-dependent RNA helicase eIF4A-3, DEAD box protein 48, Eukaryotic initiation facto, EIF4A3, DDX48, KIAA0111. UniProt: P38919. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.