Learn More
Biomatik Corporation GUCY2C anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07709
Encompass_Preferred
The GUCY2C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Heat-stable enterotoxin receptor protein (636-891AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:2000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Heat-stable enterotoxin receptor, Guanylyl cyclase C, GC-C, Intestinal guanylate cyclase, GUCY2C, GUC2C, STAR. UniProt: P25092. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7294