Learn More
Biomatik Corporation HOXA2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07710
Encompass_Preferred
The HOXA2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Homeobox protein Hox-A2 protein (26-124AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:2000-1:5000, IHC:1:20-1:200. Alternative Names: Homeobox protein Hox-A2, Homeobox protein Hox-1K, HOXA2, HOX1K. UniProt: O43364. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7295