Learn More
Biomatik Corporation MLYCD anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07715
Encompass_Preferred
Mlycd Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MLYCD. The Mlycd Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Malonyl-Coa Decarboxylase, Mitochondrial Protein (40-493Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: MLYCD. Accession Number: O95822. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7300