Learn More
Biomatik Corporation PREX2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507321
Encompass_Preferred
Prex2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PREX2. The Prex2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Phosphatidylinositol 3,4,5-Trisphosphate-Dependent Rac Exchanger 2 Protein (1417-1592Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PREX2. Accession Number: Q70Z35. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7321 Supplier Biomatik Corporation Supplier No. CAC07736
Prex2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PREX2. The Prex2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Phosphatidylinositol 3,4,5-Trisphosphate-Dependent Rac Exchanger 2 Protein (1417-1592Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PREX2. Accession Number: Q70Z35. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.