Learn More
Biomatik Corporation CAPZA1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07763
Encompass_Preferred
The CAPZA1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human F-actin-capping protein subunit alpha-1 protein (211-252AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: F-actin-capping protein subunit alpha-1, CapZ alpha-1, CAPZA1. UniProt: P52907. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7348