Learn More
Biomatik Corporation HPSE anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07770
Encompass_Preferred
The HPSE Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Heparanase protein (263-523AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Heparanase, Endo-glucoronidase, Heparanase-1, Hpa1, Heparanase 8 kDa subunit, Heparanase 50 k, HPSE, HEP, HPA, HPA1, HPR1, HPSE1, HSE1. UniProt: Q9Y251. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7355