Learn More
Biomatik Corporation IL4 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507357
Encompass_Preferred
Il4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IL4. The Il4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-4 Protein (25-153Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: IL4. Accession Number: P05112. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7357 Supplier Biomatik Corporation Supplier No. CAC07772
Il4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IL4. The Il4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-4 Protein (25-153Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: IL4. Accession Number: P05112. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.