Learn More
Biomatik Corporation MX1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507362
Encompass_Preferred
Mx1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MX1. The Mx1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon-Induced Gtp-Binding Protein Mx1 Protein (7-172Aa). Applications: ELISA, WB, IHC, IF, IP. Target Synonyms: MX1. Accession Number: P20591. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7362 Supplier Biomatik Corporation Supplier No. CAC07777
Mx1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MX1. The Mx1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon-Induced Gtp-Binding Protein Mx1 Protein (7-172Aa). Applications: ELISA, WB, IHC, IF, IP. Target Synonyms: MX1. Accession Number: P20591. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.