Learn More
Biomatik Corporation ZRSR2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507376
Encompass_Preferred
The ZRSR2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2 protein (191-372AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2, CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2, Renal carcinoma antigen NY-REN-20, U2 (RN, ZRSR2, U2AF1-RS2, U2AF1L2, U2AF1RS2, URP. UniProt: Q15696. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7376 Supplier Biomatik Corporation Supplier No. CAC07791
The ZRSR2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2 protein (191-372AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2, CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2, Renal carcinoma antigen NY-REN-20, U2 (RN, ZRSR2, U2AF1-RS2, U2AF1L2, U2AF1RS2, URP. UniProt: Q15696. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.