Learn More
Biomatik Corporation BIRC3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07793
Encompass_Preferred
The BIRC3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Baculoviral IAP repeat-containing protein 3 protein (76-245AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Baculoviral IAP repeat-containing protein 3, Apoptosis inhibitor 2, API2, Cellular inhibitor of apoptosis 2, C-IAP21, BIRC3, API2, MIHC, RNF49. UniProt: Q13489. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7378