Learn More
Biomatik Corporation AAK1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07794
Encompass_Preferred
The AAK1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human AP2-associated protein kinase 1 protein (274-465AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: AP2-associated protein kinase 1, Adaptor-associated kinase 1, AAK1, KIAA1048. UniProt: Q2M2I8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7379