Learn More
Biomatik Corporation PHLDB2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07799
Encompass_Preferred
Phldb2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PHLDB2. The Phldb2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Pleckstrin Homology-Like Domain Family B Member 2 Protein (734-1030Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PHLDB2. Accession Number: Q86SQ0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7384