Learn More
Biomatik Corporation SLC37A2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507385
Encompass_Preferred
Slc37A2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC37A2. The Slc37A2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Glucose-6-Phosphate Exchanger Slc37A2 Protein (46-84Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC37A2. Accession Number: Q8TED4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7385 Supplier Biomatik Corporation Supplier No. CAC07800
Slc37A2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC37A2. The Slc37A2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Glucose-6-Phosphate Exchanger Slc37A2 Protein (46-84Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC37A2. Accession Number: Q8TED4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.