Learn More
Biomatik Corporation BBS1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07802
Encompass_Preferred
The BBS1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Bardet-Biedl syndrome 1 protein (217-417AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Bardet-Biedl syndrome 1 protein, BBS2-like protein 2, BBS1, BBS2L2. UniProt: Q8NFJ9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7387