Learn More
Biomatik Corporation DGAT2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507388
Encompass_Preferred
The DGAT2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Diacylglycerol O-acyltransferase 2 protein (1-66AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Diacylglycerol O-acyltransferase 2, Acyl-CoA retinol O-fatty-acyltransferase (EC:2.3.1.76), ARAT, Retinol O-fatty-acyltransferase, Diglyceride acyltransfe, DGAT2, HMFN1045, UNQ738/PRO1433. UniProt: Q96PD7. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7388 Supplier Biomatik Corporation Supplier No. CAC07803
The DGAT2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Diacylglycerol O-acyltransferase 2 protein (1-66AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Diacylglycerol O-acyltransferase 2, Acyl-CoA retinol O-fatty-acyltransferase (EC:2.3.1.76), ARAT, Retinol O-fatty-acyltransferase, Diglyceride acyltransfe, DGAT2, HMFN1045, UNQ738/PRO1433. UniProt: Q96PD7. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.