Learn More
Biomatik Corporation AJAP1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07810
Encompass_Preferred
The AJAP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Adherens junction-associated protein 1 protein (308-394AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Adherens junction-associated protein 1, Membrane protein shrew-1, AJAP1, MOT8, SHREW1. UniProt: Q9UKB5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7395