Learn More
Biomatik Corporation DMRT1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07812
Encompass_Preferred
The DMRT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Doublesex- and mab-3-related transcription factor 1 protein (195-363AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Doublesex- and mab-3-related transcription factor 1, DM domain expressed in testis protein 1, DMRT1, DMT1. UniProt: Q9Y5R6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7397