Learn More
Biomatik Corporation CERK anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07817
Encompass_Preferred
The CERK Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Ceramide kinase protein (212-478AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Ceramide kinase, hCERK, Acylsphingosine kinase, Lipid kinase 4, LK4, CERK, KIAA1646. UniProt: Q8TCT0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7402