Learn More
Biomatik Corporation GBP2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507405
Encompass_Preferred
The GBP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Guanylate-binding protein 2 protein (100-259AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Guanylate-binding protein 2, GTP-binding protein 2, GBP-2, HuGBP-2, Guanine nucleotide-binding protein 2, Interferon-induced guanylate-binding protein 2, GBP2. UniProt: P32456. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7405 Supplier Biomatik Corporation Supplier No. CAC07820
The GBP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Guanylate-binding protein 2 protein (100-259AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Guanylate-binding protein 2, GTP-binding protein 2, GBP-2, HuGBP-2, Guanine nucleotide-binding protein 2, Interferon-induced guanylate-binding protein 2, GBP2. UniProt: P32456. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.