Learn More
Biomatik Corporation GFRA1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507406
Encompass_Preferred
The GFRA1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human GDNF family receptor alpha-1 protein (230-385AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: GDNF family receptor alpha-1, GDNF receptor alpha-1, GDNFR-alpha-1, GFR-alpha-1, RET ligand 1, TGF-beta-related neurotrophic factor receptor 1, GFRA1, GDNFRA, RETL1, TRNR1. UniProt: P56159. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7406 Supplier Biomatik Corporation Supplier No. CAC07821
The GFRA1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human GDNF family receptor alpha-1 protein (230-385AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: GDNF family receptor alpha-1, GDNF receptor alpha-1, GDNFR-alpha-1, GFR-alpha-1, RET ligand 1, TGF-beta-related neurotrophic factor receptor 1, GFRA1, GDNFRA, RETL1, TRNR1. UniProt: P56159. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.