Learn More
Biomatik Corporation HMGB1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07822
Encompass_Preferred
The HMGB1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human High mobility group protein B1 protein (2-215AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: High mobility group protein B1, High mobility group protein 1, HMG-1, HMGB1, HMG1. UniProt: P09429. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7407