Learn More
Biomatik Corporation SNX9 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507421
Encompass_Preferred
Snx9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SNX9. The Snx9 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sorting Nexin-9 Protein (85-200Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SNX9. Accession Number: Q9Y5X1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7421 Supplier Biomatik Corporation Supplier No. CAC07836
Snx9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SNX9. The Snx9 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sorting Nexin-9 Protein (85-200Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SNX9. Accession Number: Q9Y5X1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.