Learn More
Biomatik Corporation HRH3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07837
Encompass_Preferred
The HRH3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Histamine H3 receptor protein (236-332AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Histamine H3 receptor, H3R, HH3R, G-protein coupled receptor 97, HRH3, GPCR97. UniProt: Q9Y5N1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7422