Learn More
Biomatik Corporation AOAH anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07839
Encompass_Preferred
The AOAH Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Acyloxyacyl hydrolase protein (180-254AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Acyloxyacyl hydrolase, Acyloxyacyl hydrolase small subunit, Acyloxyacyl hydrolase large subunit, AOAH. UniProt: P28039. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7424