Learn More
Biomatik Corporation DOK3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07850
Encompass_Preferred
The DOK3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Docking protein 3 protein (1-146AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Docking protein 3, Downstream of tyrosine kinase 3, DOK3. UniProt: Q7L591. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7435