Learn More
Biomatik Corporation GABRB3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07857
Encompass_Preferred
The GABRB3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Gamma-aminobutyric acid receptor subunit beta-3 protein (328-450AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Gamma-aminobutyric acid receptor subunit beta-3, GABA (A) receptor subunit beta-3, GABRB3. UniProt: P28472. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7442