Learn More
Biomatik Corporation GCM2 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07859
Encompass_Preferred
The GCM2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Chorion-specific transcription factor GCMb protein (156-451AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: Chorion-specific transcription factor GCMb, hGCMb, GCM motif protein 2, Glial cells missing homolog 2, GCM2, GCMB. UniProt: O75603. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7444