Learn More
Biomatik Corporation GJA8 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507445
Encompass_Preferred
The GJA8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Gap junction alpha-8 protein (286-432AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Gap junction alpha-8 protein, Connexin-50, Cx50, Lens fiber protein MP70, GJA8. UniProt: P48165. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7445 Supplier Biomatik Corporation Supplier No. CAC07860
The GJA8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Gap junction alpha-8 protein (286-432AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Gap junction alpha-8 protein, Connexin-50, Cx50, Lens fiber protein MP70, GJA8. UniProt: P48165. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.