Learn More
Biomatik Corporation HSD3B1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07863
Encompass_Preferred
The HSD3B1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1 protein (127-267AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200. Alternative Names: 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3-beta-HSD I, Trophoblast antigen FDO161G, 3-beta-hydroxy-Delta (5)-steroid dehydrogenase, 3-beta-hydroxy-5-ene steroid dehydrogenase, Progesterone reductase, Steroid Delta-isomerase, Delta-5-3-ketosteroid isomerase, HSD3B1, 3BH, HSDB3A. UniProt: P14060. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7448