Learn More
Biomatik Corporation ONECUT2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07875
Encompass_Preferred
Onecut2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ONECUT2. The Onecut2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human One Cut Domain Family Member 2 Protein (214-331Aa). Applications: ELISA, WB, IHC. Target Synonyms: ONECUT2. Accession Number: O95948. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7460