Learn More
Biomatik Corporation PECAM1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07879
Encompass_Preferred
Pecam1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PECAM1. The Pecam1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Platelet Endothelial Cell Adhesion Molecule Protein (525-586Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PECAM1. Accession Number: P16284. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7464